3-(Dimethylamino)-2-(2-thienyl)acrylonitrile 95%

Lieferant: Apollo Scientific

OR32006-1G OR32006-25G OR32006-5G
APOSOR32006-1GEA 122 EUR
APOSOR32006-1G APOSOR32006-25G APOSOR32006-5G
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile 95%
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile
Formel: C₉H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00138239
CAS-Nummer: 165404-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR