3-(Dimethylamino)-2-(3-thienyl)acrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: (2Z)-3-(Dimethylamino)-2-(thiophen-3-yl)prop-2-enenitrile

OR32007-1G OR32007-25G OR32007-5G OR32007-10G
APOSOR32007-1GEA 180 EUR
APOSOR32007-1G APOSOR32007-25G APOSOR32007-5G APOSOR32007-10G
3-(Dimethylamino)-2-(3-thienyl)acrylonitrile 95%
3-(Dimethylamino)-2-(3-thienyl)acrylonitrile
Formel: C₉H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00127051
CAS-Nummer: 165404-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR