N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide 95%

Lieferant: Apollo Scientific

OR32008-1G
APOSOR32008-1GEA 1570 EUR
APOSOR32008-1G
N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide 95%
N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide
Formel: C₁₄H₁₇N₃O₂
Molecular Weight: 259.304 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139095
CAS-Nummer: 241127-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR