(3E)-1-(2-Hydroxyphenyl)-3-(methoxyimino)propan-1-one 95%

Lieferant: Apollo Scientific

OR32011-1G
APOSOR32011-1GEA 1570 EUR
APOSOR32011-1G
(3E)-1-(2-Hydroxyphenyl)-3-(methoxyimino)propan-1-one 95%
(3E)-1-(2-Hydroxyphenyl)-3-(methoxyimino)propan-1-one
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127059
CAS-Nummer: 89836-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR