1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one 95%
Lieferant: Apollo Scientific
OR32012-1G
APOSOR32012-1GEA
1570
EUR
APOSOR32012-1G
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one 95%
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one
Formel:
C₁₆H₁₆N₂O₂ Molecular Weight: 268,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139309 CAS-Nummer: 241488-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...