1-[4-(1H-Pyrazol-5-yl)phenyl]-1,4-dihydropyridin-4-one 95%

Lieferant: Apollo Scientific

OR32013-1G
APOSOR32013-1GEA 1570 EUR
APOSOR32013-1G
1-[4-(1H-Pyrazol-5-yl)phenyl]-1,4-dihydropyridin-4-one 95%
1-[4-(1H-Pyrazol-5-yl)phenyl]-1,4-dihydropyridin-4-one
Formel: C₁₄H₁₁N₃O
Molecular Weight: 237,257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139310
CAS-Nummer: 241488-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR