(3Z)-N-(4-Chlorophenyl)-3-(methoxyimino)propanamide 95%
Lieferant: Apollo Scientific
OR32017-1G
APOSOR32017-1GEA
1570
EUR
APOSOR32017-1G
(3Z)-N-(4-Chlorophenyl)-3-(methoxyimino)propanamide 95%
(3Z)-N-(4-Chlorophenyl)-3-(methoxyimino)propanamide
Formel:
C₁₀H₁₁ClN₂O₂ Molecular Weight: 226,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00201878 CAS-Nummer: 241132-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...