(2E)-3-(Dimethylamino)-N-(4-phenoxyphenyl)prop-2-enamide 95%

Lieferant: Apollo Scientific

OR32018-1G
APOSOR32018-1GEA 1570 EUR
APOSOR32018-1G
(2E)-3-(Dimethylamino)-N-(4-phenoxyphenyl)prop-2-enamide 95%
(2E)-3-(Dimethylamino)-N-(4-phenoxyphenyl)prop-2-enamide
Formel: C₁₇H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00201884
CAS-Nummer: 241132-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR