{Hexahydro-2H-[1,2]oxazolo[2,3-a]pyridin-2-yl}methanol 95%
Lieferant: Apollo Scientific
OR32019-1G
APOSOR32019-1GEA
1570
EUR
APOSOR32019-1G
{Hexahydro-2H-[1,2]oxazolo[2,3-a]pyridin-2-yl}methanol 95%
{Hexahydro-2H-[1,2]oxazolo[2,3-a]pyridin-2-yl}methanol
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140539 CAS-Nummer: 241132-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...