3-[(E)-2-(Dimethylamino)ethenyl]-5,5-dimethylcyclohex-2-en-1-one 95%

Lieferant: Apollo Scientific

OR32021-1G
APOSOR32021-1GEA 1570 EUR
APOSOR32021-1G
3-[(E)-2-(Dimethylamino)ethenyl]-5,5-dimethylcyclohex-2-en-1-one 95%
3-[(E)-2-(Dimethylamino)ethenyl]-5,5-dimethylcyclohex-2-en-1-one
Formel: C₁₂H₁₉NO
Molecular Weight: 193.285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140800
CAS-Nummer: 128475-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR