N-[(7E)-9,9-Dimethyl-1-oxa-2-azaspiro[4.5]dec-2-en-7-ylidene]hydroxylamine 95%

Lieferant: Apollo Scientific

OR32022-1G
APOSOR32022-1GEA 1570 EUR
APOSOR32022-1G
N-[(7E)-9,9-Dimethyl-1-oxa-2-azaspiro[4.5]dec-2-en-7-ylidene]hydroxylamine 95%
N-[(7E)-9,9-Dimethyl-1-oxa-2-azaspiro[4.5]dec-2-en-7-ylidene]hydroxylamine
Formel: C₁₀H₁₆N₂O₂
Molecular Weight: 196,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140801
CAS-Nummer: 128475-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR