Ethyl-1H-1,2,4-triazol-1-ylacetate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(1H-1,2,4-triazol-1-yl)acetate

OR32023-1G
APOSOR32023-1GEA 1330 EUR
APOSOR32023-1G
Ethyl-1H-1,2,4-triazol-1-ylacetate 95%
Ethyl-1H-1,2,4-triazol-1-ylacetate
Formel: C₆H₉N₃O₂
Molecular Weight: 155,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215199
CAS-Nummer: 56563-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR