(2Z)-3-(Dimethylamino)-1-phenyl-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one 95%

Lieferant: Apollo Scientific

OR32024-1G
APOSOR32024-1GEA 1570 EUR
APOSOR32024-1G
(2Z)-3-(Dimethylamino)-1-phenyl-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one 95%
(2Z)-3-(Dimethylamino)-1-phenyl-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one
Formel: C₁₃H₁₄N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00214690
CAS-Nummer: 241146-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR