1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole 95%

Lieferant: Apollo Scientific

OR32025-1G
APOSOR32025-1GEA 1570 EUR
APOSOR32025-1G
1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole 95%
1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole
Formel: C₁₁H₉N₅
Molecular Weight: 211,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214691
CAS-Nummer: 241146-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR