5,5-Dimethyl-2-(2-oxopropyl)cyclohexane-1,3-dione 95%
Lieferant: Apollo Scientific
OR32031-1G
APOSOR32031-1GEA
1120
EUR
APOSOR32031-1G
5,5-Dimethyl-2-(2-oxopropyl)cyclohexane-1,3-dione 95%
5,5-Dimethyl-2-(2-oxopropyl)cyclohexane-1,3-dione
Formel:
C₁₁H₁₆O₃ Molecular Weight: 196,243 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793015 CAS-Nummer: 13148-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...