6-Chloro-2-(4-methylphenyl)-1H-1,3-benzodiazol-1-ol 95%

Lieferant: Apollo Scientific

OR32033-1G
APOSOR32033-1GEA 1570 EUR
APOSOR32033-1G
6-Chloro-2-(4-methylphenyl)-1H-1,3-benzodiazol-1-ol 95%
6-Chloro-2-(4-methylphenyl)-1H-1,3-benzodiazol-1-ol
Formel: C₁₄H₁₁ClN₂O
Molecular Weight: 258,703 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141397
CAS-Nummer: 250610-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR