2-Chloro-6-(methylsulphanyl)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-(methylthio)pyridine

OR32036-1G
APOSOR32036-1GEA 874 EUR
APOSOR32036-1G
2-Chloro-6-(methylsulphanyl)pyridine 95%
2-Chloro-6-(methylsulphanyl)pyridine
Formel: C₆H₆ClNS
Molecular Weight: 159,637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173422
CAS-Nummer: 77145-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR