1-Bromo-4-isobutylbenzene

Lieferant: Apollo Scientific
Achtung

OR3204-25G
APOSOR3204-25GEA 89.9 EUR
APOSOR3204-25G
1-Bromo-4-isobutylbenzene
1-Bromo-4-isobutylbenzene
Formel: C₁₀H₁₃Br
Molecular Weight: 213.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425886
CAS-Nummer: 2051-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR