2-Chloro-N-[4-(piperidin-1-yl)phenyl]acetamide 95%

Lieferant: Apollo Scientific

OR32040-1G
APOSOR32040-1GEA 1570 EUR
APOSOR32040-1G
2-Chloro-N-[4-(piperidin-1-yl)phenyl]acetamide 95%
2-Chloro-N-[4-(piperidin-1-yl)phenyl]acetamide
Formel: C₁₃H₁₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03119623
CAS-Nummer: 379255-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR