2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]propanamide 95%

Lieferant: Apollo Scientific

OR32042-1G
APOSOR32042-1GEA 336 EUR
APOSOR32042-1G
2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]propanamide 95%
2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]propanamide
Formel: C₁₃H₁₇ClN₂O
Molecular Weight: 252,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173029
CAS-Nummer: 251097-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR