1-(3-Chloro-4-methoxyphenyl)-3-(propan-2-yl)urea, tech

Lieferant: Apollo Scientific

OR32050-1G
APOSOR32050-1GEA 1570 EUR
APOSOR32050-1G
1-(3-Chloro-4-methoxyphenyl)-3-(propan-2-yl)urea, tech
1-(3-Chloro-4-methoxyphenyl)-3-(propan-2-yl)urea
Formel: C₁₁H₁₅ClN₂O₂
Molecular Weight: 242,702 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316321
CAS-Nummer: 252058-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR