1-(3-Chloro-4-methoxyphenyl)-3-(prop-2-en-1-yl)urea, tech

Lieferant: Apollo Scientific

OR32052-1G
APOSOR32052-1GEA 1570 EUR
APOSOR32052-1G
1-(3-Chloro-4-methoxyphenyl)-3-(prop-2-en-1-yl)urea, tech
1-(3-Chloro-4-methoxyphenyl)-3-(prop-2-en-1-yl)urea
Formel: C₁₁H₁₃ClN₂O₂
Molecular Weight: 240,686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124019
CAS-Nummer: 860609-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR