6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32053-1G
APOSOR32053-1GEA 1570 EUR
APOSOR32053-1G
6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₂H₆Cl₂N₂O
Molecular Weight: 265,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214977
CAS-Nummer: 142499-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR