1-{6-Phenyl-3aH,4H,6H,8H,8aH-[1,3]dioxepino[5,6-d][1,2]oxazol-3-yl}ethan-1-one, tech

Lieferant: Apollo Scientific

OR32054-1G
APOSOR32054-1GEA 1570 EUR
APOSOR32054-1G
1-{6-Phenyl-3aH,4H,6H,8H,8aH-[1,3]dioxepino[5,6-d][1,2]oxazol-3-yl}ethan-1-one, tech
1-{6-Phenyl-3aH,4H,6H,8H,8aH-[1,3]dioxepino[5,6-d][1,2]oxazol-3-yl}ethan-1-one
Formel: C₁₄H₁₅NO₄
Molecular Weight: 261,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664164
CAS-Nummer: 317820-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR