3-Oxo-N-(pyridin-2-yl)butanamide 95%

Lieferant: Apollo Scientific

OR32056-1G
APOSOR32056-1GEA 671 EUR
APOSOR32056-1G
3-Oxo-N-(pyridin-2-yl)butanamide 95%
3-Oxo-N-(pyridin-2-yl)butanamide
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178.188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023427
CAS-Nummer: 1657-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR