Methyl 4-[(2E)-3-(pyrrolidin-1-yl)but-2-enamido]benzoate, tech

Lieferant: Apollo Scientific

OR32057-1G
APOSOR32057-1GEA 1570 EUR
APOSOR32057-1G
Methyl 4-[(2E)-3-(pyrrolidin-1-yl)but-2-enamido]benzoate, tech
Methyl 4-[(2E)-3-(pyrrolidin-1-yl)but-2-enamido]benzoate
Formel: C₁₆H₂₀N₂O₃
Molecular Weight: 288,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244735
CAS-Nummer: 337921-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR