(5E)-4-Oxo-6-phenylhex-5-enoic acid, tech

Lieferant: Apollo Scientific

OR32058-1G
APOSOR32058-1GEA 1570 EUR
APOSOR32058-1G
(5E)-4-Oxo-6-phenylhex-5-enoic acid, tech
(5E)-4-Oxo-6-phenylhex-5-enoic acid
Formel: C₁₂H₁₂O₃
Molecular Weight: 204,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020542
CAS-Nummer: 121365-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR