6-[(E)-2-Phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32059-1G
APOSOR32059-1GEA 1570 EUR
APOSOR32059-1G
6-[(E)-2-Phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
6-[(E)-2-Phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793232
CAS-Nummer: 83516-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR