2-Methyl-6-[(E)-2-phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32060-1G
APOSOR32060-1GEA 1570 EUR
APOSOR32060-1G
2-Methyl-6-[(E)-2-phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
2-Methyl-6-[(E)-2-phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214.263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793234
CAS-Nummer: 1421944-09-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR