6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32061-1G
APOSOR32061-1GEA 1570 EUR
APOSOR32061-1G
6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one
Formel: C₁₅H₁₆N₂O
Molecular Weight: 240,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793235
CAS-Nummer: 1421944-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR