6-[(E)-2-Phenylethenyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR32062-1G
APOSOR32062-1GEA 1570 EUR
APOSOR32062-1G
6-[(E)-2-Phenylethenyl]pyridazin-3-ol, tech
6-[(E)-2-Phenylethenyl]pyridazin-3-ol
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793247
CAS-Nummer: 83516-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR