6-[(E)-2-(4-Methoxyphenyl)ethenyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR32064-1G
APOSOR32064-1GEA 1570 EUR
APOSOR32064-1G
6-[(E)-2-(4-Methoxyphenyl)ethenyl]pyridazin-3-ol, tech
6-[(E)-2-(4-Methoxyphenyl)ethenyl]pyridazin-3-ol
Formel: C₁₃H₁₂N₂O₂
Molecular Weight: 228,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793268
CAS-Nummer: 83516-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR