[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride, tech

Lieferant: Apollo Scientific

OR32065-1G
APOSOR32065-1GEA 1570 EUR
APOSOR32065-1G
[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride, tech
[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride
Formel: C₁₅H₁₇NO₂·ClH
Molecular Weight: 279,762 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663770
CAS-Nummer: 477708-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR