4-(4-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarbonitrile, tech

Lieferant: Apollo Scientific

OR32066-1G
APOSOR32066-1GEA 0 EUR
APOSOR32066-1G
4-(4-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarbonitrile, tech
4-(4-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarbonitrile
Formel: C₁₅H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00663771
CAS-Nummer: 67438-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR