N'-[(1E)-1-(3,4-Dichlorophenyl)ethylidene]methoxycarbohydrazide, tech
Lieferant: Apollo Scientific
OR32067-1G
APOSOR32067-1GEA
1570
EUR
APOSOR32067-1G
N'-[(1E)-1-(3,4-Dichlorophenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(3,4-Dichlorophenyl)ethylidene]methoxycarbohydrazide
Formel:
C₁₀H₁₀Cl₂N₂O₂ Molecular Weight: 261,105 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792939 CAS-Nummer: 337924-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...