N'-[(1E)-1-(3-Methoxyphenyl)ethylidene]methoxycarbohydrazide, tech
Lieferant: Apollo Scientific
OR32070-1G
APOSOR32070-1GEA
1570
EUR
APOSOR32070-1G
N'-[(1E)-1-(3-Methoxyphenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(3-Methoxyphenyl)ethylidene]methoxycarbohydrazide
Formel:
C₁₁H₁₄N₂O₃ Molecular Weight: 222,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792942 CAS-Nummer: 205121-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...