N'-[(1E)-1-(Naphthalen-2-yl)ethylidene]methoxycarbohydrazide, tech

Lieferant: Apollo Scientific

OR32071-1G
APOSOR32071-1GEA 1570 EUR
APOSOR32071-1G
N'-[(1E)-1-(Naphthalen-2-yl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(Naphthalen-2-yl)ethylidene]methoxycarbohydrazide
Formel: C₁₄H₁₄N₂O₂
Molecular Weight: 242,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00770165
CAS-Nummer: 54886-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR