N'-[(1E)-1-(4-Methoxyphenyl)ethylidene]methoxycarbohydrazide, tech

Lieferant: Apollo Scientific

OR32072-1G
APOSOR32072-1GEA 1570 EUR
APOSOR32072-1G
N'-[(1E)-1-(4-Methoxyphenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(4-Methoxyphenyl)ethylidene]methoxycarbohydrazide
Formel: C₁₁H₁₄N₂O₃
Molecular Weight: 222,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792949
CAS-Nummer: 1202248-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR