N'-[(2E,3E)-4-Phenylbut-3-en-2-ylidene]methoxycarbohydrazide, tech

Lieferant: Apollo Scientific

OR32073-1G
APOSOR32073-1GEA 1570 EUR
APOSOR32073-1G
N'-[(2E,3E)-4-Phenylbut-3-en-2-ylidene]methoxycarbohydrazide, tech
N'-[(2E,3E)-4-Phenylbut-3-en-2-ylidene]methoxycarbohydrazide
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792955
CAS-Nummer: 358723-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR