3-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile, tech

Lieferant: Apollo Scientific

OR32077-1G
APOSOR32077-1GEA 1570 EUR
APOSOR32077-1G
3-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile, tech
3-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793353
CAS-Nummer: 341965-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR