Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech

Lieferant: Apollo Scientific

OR32079-1G
APOSOR32079-1GEA 1570 EUR
APOSOR32079-1G
Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech
Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793358
CAS-Nummer: 341965-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR