N-(4-Cyanophenyl)-2-methylprop-2-enamide, tech

Lieferant: Apollo Scientific

OR32081-1G
APOSOR32081-1GEA 1570 EUR
APOSOR32081-1G
N-(4-Cyanophenyl)-2-methylprop-2-enamide, tech
N-(4-Cyanophenyl)-2-methylprop-2-enamide
Formel: C₁₁H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00793366
CAS-Nummer: 90617-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR