1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one, tech

Lieferant: Apollo Scientific

OR32083-1G
APOSOR32083-1GEA 1570 EUR
APOSOR32083-1G
1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one, tech
1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315912
CAS-Nummer: 321385-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR