Ethyl 2-[(4Z)-4,5,6,7-tetrahydro-1-benzofuran-4-ylidene]acetate, tech
Lieferant: Apollo Scientific
OR32084-1G
APOSOR32084-1GEA
1570
EUR
APOSOR32084-1G
Ethyl 2-[(4Z)-4,5,6,7-tetrahydro-1-benzofuran-4-ylidene]acetate, tech
Ethyl 2-[(4Z)-4,5,6,7-tetrahydro-1-benzofuran-4-ylidene]acetate
Formel:
C₁₂H₁₄O₃ Molecular Weight: 206,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01568614 CAS-Nummer: 273929-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...