4-Chloro-2-(2-oxopropyl)-5-(piperidin-1-yl)-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32090-1G
APOSOR32090-1GEA 1570 EUR
APOSOR32090-1G
4-Chloro-2-(2-oxopropyl)-5-(piperidin-1-yl)-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(2-oxopropyl)-5-(piperidin-1-yl)-2,3-dihydropyridazin-3-one
Formel: C₁₂H₁₆ClN₃O₂
Molecular Weight: 269,727 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124084
CAS-Nummer: 860609-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR