2-{[(4-Chlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Lieferant: Apollo Scientific
OR32092-1G
APOSOR32092-1GEA
1570
EUR
APOSOR32092-1G
2-{[(4-Chlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(4-Chlorophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel:
C₁₃H₁₂ClNO₂ Molecular Weight: 249,693 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00101155 CAS-Nummer: 62370-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...