2-{[(3,5-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32093-1G
APOSOR32093-1GEA 1570 EUR
APOSOR32093-1G
2-{[(3,5-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(3,5-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel: C₁₃H₁₁Cl₂NO₂
Molecular Weight: 284,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171109
CAS-Nummer: 55118-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR