2-{[(3,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Lieferant: Apollo Scientific
OR32094-1G
APOSOR32094-1GEA
1570
EUR
APOSOR32094-1G
2-{[(3,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(3,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel:
C₁₃H₁₁Cl₂NO₂ Molecular Weight: 284,138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569051 CAS-Nummer: 341968-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...