2-{[(2,3-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Lieferant: Apollo Scientific
OR32095-1G
APOSOR32095-1GEA
1570
EUR
APOSOR32095-1G
2-{[(2,3-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(2,3-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel:
C₁₃H₁₁Cl₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569052 CAS-Nummer: 341968-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...