2-{[(2,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32096-1G
APOSOR32096-1GEA 1570 EUR
APOSOR32096-1G
2-{[(2,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(2,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel: C₁₃H₁₁Cl₂NO₂
Molecular Weight: 284,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569053
CAS-Nummer: 341968-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR